missing translation for 'onlineSavingsMsg'
Learn More

FMOC-Cys(tBu)-OH, 98%

Catalog No. AC431270050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431270050 5 g Glass bottle
AC431270250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431270050 Supplier Thermo Scientific Chemicals Supplier No. 431270050

CAS: 67436-13-9 | C22H25NO4S | 399.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67436-13-9
Molecular Formula C22H25NO4S
Molecular Weight (g/mol) 399.51
MDL Number MFCD00037130
InChI Key IXAYZHCPEYTWHW-IBGZPJMESA-N
Synonym fmoc-cys tbu-oh, fmoc-s-tert-butyl-l-cysteine, fmoc-s-t-butyl-l-cysteine, n-fmoc-s-tert-butyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butylthio propanoic acid, fmoc-r-2-amino-3-s-t-butylthio butanoic acid, 2r-3-tert-butylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1716, pubchem19015, fmoc-cys but-oh
PubChem CID 7009544
IUPAC Name (2R)-3-tert-butylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)SCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 137.1°C to 138.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -21.0° to -23.0° (20°C, 589nm) (c=1, DMF)
Formula Weight 399.51
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material FMOC-Cys(tBu)-OH
Show More Show Less

RUO – Research Use Only