missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxamine Maleate 98.0+%, TCI America™

Catalog No. p-7129944
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08581G 1 g
F08585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08581G Supplier TCI America Supplier No. F08581G
Only null left

Chemical Identifiers

CAS 61718-82-9
Molecular Formula C19H25F3N2O6
Molecular Weight (g/mol) 434.41
MDL Number MFCD00269809,MFCD00269809
InChI Key LFMYNZPAVPMEGP-PIDGMYBPSA-N
Synonym (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone O-(2-Aminoethyl)oxime Maleate
PubChem CID 119090957
IUPAC Name (2Z)-but-2-enedioic acid; (E)-(2-aminoethoxy)({5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene})amine
SMILES OC(=O)\C=C/C(O)=O.COCCCC\C(=N/OCCN)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 434.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluvoxamine Maleate
RTECSNumber SA9230000
TSCA No
Recommended Storage Refrigerator