missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluvoxamine Maleate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 61718-82-9 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C15H21F3N2O2 · C4H4O4 |
| MDL Number | MFCD00269809 |
| Synonym | Fluvoxamine maleate; (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 434.41 |
| Grade | Certified Reference Material |