missing translation for 'onlineSavingsMsg'
Learn More

Flutamide 98.0+%, TCI America™

Catalog No. p-7129820
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F066325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F066325G Supplier TCI America Supplier No. F066325G

Chemical Identifiers

CAS 13311-84-7
Molecular Formula C11H11F3N2O3
Molecular Weight (g/mol) 276.22
MDL Number MFCD00072009
InChI Key MKXKFYHWDHIYRV-UHFFFAOYSA-N
Synonym flutamide, eulexin, niftolide, niftholide, nfba, drogenil, 2-methyl-n-4-nitro-3-trifluoromethyl phenyl propanamide, niftolid, flutamidum, flutamida
PubChem CID 3397
ChEBI CHEBI:5132
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Melting Point 112°C
Quantity 25 g
Formula Weight 276.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Flutamide
RTECSNumber UG5700000
TSCA No