missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flutamide

$177.60 - $177.60

Chemical Identifiers

CAS 13311-84-7
Molecular Formula C11H11F3N2O3
Molecular Weight (g/mol) 276.22
MDL Number MFCD00072009
InChI Key MKXKFYHWDHIYRV-UHFFFAOYSA-N
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466450050
View Documents
Thermo Scientific Chemicals
466450050
5 g
Each for $177.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13311-84-7
276.22
MKXKFYHWDHIYRV-UHFFFAOYSA-N
CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
C11H11F3N2O3
MFCD00072009
2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide

Specifications

FLUTAMIDE
110.0°C to 114.0°C
Conforms
C11H11F3N2O3
5 g
MKXKFYHWDHIYRV-UHFFFAOYSA-N
2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
276.21
Flutamide
13311-84-7
White to Yellow
glass bottle
MFCD00072009
insoluble aq. soln.
CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
276.22
Powder
Videos
Safety and Handling

Safety and Handling

GHS08: Health hazard

GHS07: Harmful

ShelfLife : 5 years

storageNote1 : normal conditions

SDS
Documents

Documents

RUO – Research Use Only