missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flutamide

Catalog No. AC466450050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466450050 5 g
AC466450250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466450050 Supplier Thermo Scientific Chemicals Supplier No. 466450050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13311-84-7
Molecular Formula C11H11F3N2O3
Molecular Weight (g/mol) 276.22
MDL Number MFCD00072009
InChI Key MKXKFYHWDHIYRV-UHFFFAOYSA-N
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Alpha Vector FLUTAMIDE
Melting Point 110.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
Solubility Information insoluble aq. soln.
Formula Weight 276.21
Physical Form Powder
Chemical Name or Material Flutamide
Hazard Category GHS07: Harmful
Hazard Statement GHS08: Health hazard
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only