missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7129874
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07501G 1 g
F07505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07501G Supplier TCI America Supplier No. F07501G
Only null left

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO
Molecular Weight (g/mol) 345.79
MDL Number MFCD00214288
InChI Key GIYXAJPCNFJEHY-UHFFFAOYSA-N
Synonym fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
PubChem CID 62857
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl

Specifications

Melting Point 159°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 345.79
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluoxetine Hydrochloride
RTECSNumber UI4050000
TSCA No