missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019604
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019604 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019604 Supplier Merck Emd Millipore Supplier No. PHR13941G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Fluoxetine Hydrochloride is a selective serotonin-reuptake inhibitor used as an antidepressant. It is also utilized for the treatment of obsessive-compulsive disorders. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 56296-78-7
Physical Form Neat
Linear Formula C17H18F3 NO · HCl
MDL Number MFCD00214288
Synonym Fluoxetine hydrochloride; (+/-)-N-Methyl-gamma-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride; LY-110,140 hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 345.79
Grade Certified Reference Material

Sold under license from Eli Lilly and Company.