missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111018498
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018498 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018498 Supplier Merck Emd Millipore Supplier No. 3401210MGR

Fluoxetine hydrochloride is an antidepressant drug and a selective serotonin reuptake inhibitor, which is widely used for the treatment of major depressive disorders, obsessive-compulsive disorder and panic fits.

Pharmaceutical (small molecule)

Specifications

CAS 56296-78-7
Physical Form Neat
Linear Formula C17H18F3 NO · HCl
MDL Number MFCD00214288
Synonym (+/-)-N-Methyl-gamma-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride; LY-110,140 hydrochloride; Prozac™
Type Analytical Standard
Quantity 10 mg
Formula Weight 345.79
Grade Analytical Standard

Sold under license from Eli Lilly and Company.