missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine hydrochloride, 99%

A selective serotonin reuptake inhibitor | CAS: 56296-78-7 | C17H19ClF3NO | 345.79 g/mol

$328.34 - $916.19

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO
Molecular Weight (g/mol) 345.79
MDL Number MFCD00214288
InChI Key GIYXAJPCNFJEHY-UHFFFAOYSA-N
Synonym fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
PubChem CID 62857
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61197MF
View Documents
Thermo Scientific Chemicals
J61197MF
50 mg
Each for $328.34
Only null left
 
AAJ61197MD
View Documents
Thermo Scientific Chemicals
J61197MD
250 mg
Each for $916.19
Only null left
 
Description

Description

Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

56296-78-7
345.79
GIYXAJPCNFJEHY-UHFFFAOYSA-N
62857
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
C17H19ClF3NO
MFCD00214288
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride

Specifications

56296-78-7
White
MFCD00214288
14,4185
Soluble in dimethylsulfoxide and water.
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
345.79
345.79
Powder
158°C to 159°C
C17H19ClF3NO
50 mg
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
62857
99%
Fluoxetine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c

H302-H315-H318

EINECSNumber : 260-101-2

RTECSNumber : UI4050000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only