missing translation for 'onlineSavingsMsg'
Learn More

Fluosilicic Acid, 25%, Spectrum™ Chemical

Catalog No. 18610632
Change view
Click to view available options
Quantity:
500 g
200 L
Packaging:
Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610632 500 g Poly Bottle
18610631 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610632 Supplier Spectrum Chemical Mfg Cor Supplier No. FL130500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16961-83-4 , 7732-18-5
Molecular Weight (g/mol) 144.08

Specifications

CAS Max % 20 to 40%, 60 to 80%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 19 to 20.6%
Grade Reagent
Chemical Name or Material Fluosilicic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.