missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader

Catalog No. 502576101
Encompass
Change view
Click to view available options
Excitation/Emission:
334/450 nm
390/450 nm
390/513 nm
452/545 nm
460 nm, 618 nm (Donor/Acceptor)
497/542 nm
535/595 nm
625/677 nm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Excitation/Emission
502576101 452/545 nm
502576104 497/542 nm
502576107 535/595 nm
502576106 625/677 nm
502576103 390/513 nm
502576102 390/450 nm
502576100 334/450 nm
502576105 460 nm, 618 nm (Donor/Acceptor)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 502576101 Supplier Thermo Scientific™ Supplier No. VAPAI011

Varioskan ALF Multimode Microplate Reader Filter Sets are high quality, validated, and available for various fluorometric and luminometric applications.

With the Varioskan ALF Multimode Microplate Reader, fluorescence and certain luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Quant-iT DNA quantitation assays and PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter pair positions are available for customized fluorometric and luminometric applications such as measurement of various Alexa Fluor™ and other common dyes or Promega NanoBRET™ assays. The filters are available as predefined, carefully tested, and validated filter sets, to ensure the optimal performance of selected assays.

Varioskan ALF instrument includes the following factory installed fluorometric filter pairs:

  • Coumarin family filter pair
    • Excitation 345 nm, emission 450 nm
  • Fluorescein family filter pair
    • Excitation 485, emission 525 nm
  • Resorufin family filter pair
    • Excitation 555 nm, emission 615 nm

In addition to factory installed filters, a range of validated filter sets are available for various fluorometric and luminometric applications.

  • Filter sets consists of high transmission filters for high excitation and emission efficiency.
  • Filter sets are designed to efficiently block unwanted light as well as unwanted emitted light to ensure high signal and low background.
  • Varioskan ALF filter installation is effortless
    • The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software.
    • SkanIt software-controlled filter installation process prevents accidental mistakes ensuring error-free

Specifications

Product Type Filter Set
For Use With (Equipment) Varioskan ALF Multimode Microplate Reader
Plate Type 6 to 384-well microplates
Excitation/Emission 452/545 nm
Quantity Each
For Use With Alexa Fluor™ 430 and equivalent dyes

For Laboratory Use