missing translation for 'onlineSavingsMsg'
Learn More

Fluoroboric Acid, 50% (Laboratory), Fisher Chemical

Catalog No. A188500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A188500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A188500 Supplier Thermo Fisher Scientific Supplier No. FLA188500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16872-11-0 , 7732-18-5 , 10043-35-3
Molecular Formula BF4H
Molecular Weight (g/mol) 87.812
MDL Number MFCD00011345
InChI Key ODGCEQLVLXJUCC-UHFFFAOYSA-O
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
PubChem CID 28118
ChEBI CHEBI:38902
IUPAC Name hydron;tetrafluoroborate
SMILES [H+].[B-](F)(F)(F)F

Specifications

Color Yellow
Melting Point -90°C
Boiling Point 130°C
Density 1.41g/cm³
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 5.1mmHg at 20°C
Assay Percent Range 48 to 50%
Quantity 500 mL
Physical Form Liquid
UN Number 1775
Formula Weight 87.81
Percent Purity 48 to 50%
Grade Laboratory
Chemical Name or Material Fluoroboric Acid, 50%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive