missing translation for 'onlineSavingsMsg'
Learn More

Fluoro(phenylthio)acetonitrile 97.0+%, TCI America™

Catalog No. F0464100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0464100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0464100MG Supplier TCI America Supplier No. F0464100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130612-84-9
Molecular Formula C8H6FNS
Molecular Weight (g/mol) 167.20
MDL Number MFCD06797105
InChI Key VAMSBCYZRBSCPJ-UHFFFAOYNA-N
PubChem CID 15634483
IUPAC Name 2-fluoro-2-(phenylsulfanyl)acetonitrile
SMILES FC(SC1=CC=CC=C1)C#N

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 167.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluoro(phenylthio)acetonitrile