missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-tetramethylformamidinium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. F07261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07261G 1 g
F07265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07261G Supplier TCI America Supplier No. F07261G

Chemical Identifiers

CAS 164298-23-1
Molecular Formula C5H12F7N2P
Molecular Weight (g/mol) 264.128
MDL Number MFCD02684443
InChI Key ZAVXOOLKAGPJPI-UHFFFAOYSA-N
Synonym tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate
PubChem CID 2774761
IUPAC Name [dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 1 g
Formula Weight 264.13
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Fluoro-N,N,N′,N′-tetramethylformamidinium Hexafluorophosphate