missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%

Catalog No. p-7036595
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5491003 1 g
AAH5491006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5491003 Supplier Thermo Scientific Chemicals Supplier No. H5491003
Only null left

CAS: 164298-23-1 | C5H12F7N2P | 264.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 164298-23-1
Molecular Formula C5H12F7N2P
Molecular Weight (g/mol) 264.128
MDL Number MFCD02684443
InChI Key ZAVXOOLKAGPJPI-UHFFFAOYSA-N
Synonym tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate
PubChem CID 2774761
IUPAC Name [dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 104°C to 111°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 264.12
Percent Purity 98%
Chemical Name or Material Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only