missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 1000 ppm F-, Ricca Chemical

Supplier:  Ricca Chemical Company 317316

 View more versions of this product

Catalog No. 317316


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Fluoride Standard
7681-49-4,7732-18-5
1000ppm F-
1mL = 1mg F-
FNa
Colorless
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
500 mL
CHEBI:28741
Natural Poly Bottle
98.0,0.22
100.0,0.23
Liquid
MFCD00003524
100°C
[F-].[Na+]
41.99
5235

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
FNa
MFCD00003524
5235
sodium fluoride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions