missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorexon

Catalog No. AC410630050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410630050 5 g Glass bottle
AC410630250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC410630050 Supplier Thermo Scientific Chemicals Supplier No. 410630050
Only null left

CAS: 1461-15-0 | C30H26N2O13 | 622.539 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
MDL Number MFCD00005049
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Bis[N, N-bis(carboxymethyl)aminomethyl]fluorescein, Calcein, Fluorescein Complexon
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Infrared Spectrum Authentic
Formula Weight 622.54
Packaging Glass bottle
Suitability Suitable for calcium determination
UV Spectrometry Complies
Color Brown/Orange to Orange/Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Fluorexon
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-957-1
TSCA TSCA

RUO – Research Use Only