missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum Fluorescent Lamp Recycling Drum: PROMO
Youngstown Barrel & Drum Fluorescent Lamp Recycling Drum are constructed of sturdy fiberboard and are an excellent solution for fluorescent lamp recycling.
Supplier: Youngstown Barrel & Drum 2064PMO
Description
- Open-head fiber drums for the transport and storage involved in recycling fluorescent lamps.
- Available in 16 x 76 in., 16 x 49 in., and 24 x 49 in. dimensions.
Specifications
| Fiber Drum | |
| Fiber | |
| 96 in. | |
| 15 lb. |
| Fluorescent Lamp Recycling | |
| Open | |
| 16 in. |