missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagents
Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$134.11 - $440.96
Chemical Identifiers
| CAS | 518-47-8 |
|---|---|
| Molecular Formula | C20H10Na2O5 |
| Molecular Weight (g/mol) | 376.28 |
| MDL Number | MFCD00167039 |
| InChI Key | NJDNXYGOVLYJHP-UHFFFAOYSA-L |
| Synonym | Disodium Fluorescein, Uranine, Color Index 45350 |
| IUPAC Name | disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
Chemical Identifiers
| 518-47-8 | |
| 376.28 | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| C20H10Na2O5 | |
| MFCD00167039 | |
| Disodium Fluorescein, Uranine, Color Index 45350 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
Specifications
| 518-47-8 | |
| MFCD00167039 | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
| Reagent | |
| 1 | |
| Solid |
| C20H10Na2O5 | |
| Disodium Fluorescein, Uranine, Color Index 45350 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 | |
| 376.28 | |
| Glass Bottle | |
| Orange to Red | |
| Fluorescein, Water Soluble CI 45350 (Acid Yellow 73, Sodium Fluorescein) |