missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Water Soluble CI 45350 (Acid Yellow 73; Sodium Fluorescein), Reagents
Specifications
Specifications
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| Color | Orange to Red |
| Quantity | 100 g |
| Physical Form | Solid |
| Chemical Name or Material | Fluorescein, Water Soluble CI 45350 (Acid Yellow 73, Sodium Fluorescein) |