missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein sodium salt, ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014420
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014420 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014420 Supplier Merck Emd Millipore Supplier No. 30181100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 518-47-8
Physical Form Neat
Linear Formula C20H10 Na2O5
MDL Number MFCD00167039
Synonym Acid Yellow 73; D&C;Yellow No. 8; NaFl; NaFluo; Sodium fluorescein; Uranine
Type Analytical Standard
Quantity 100 mg
Formula Weight 376.27
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard