missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein Isothiocyanate Isomer I, 90%, Pure

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol

$137.85 - $1,769.81

Chemical Identifiers

CAS 3326-32-7
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.381
MDL Number MFCD00005063
InChI Key MHMNJMPURVTYEJ-UHFFFAOYSA-N
Synonym 5-FITC, 5-Isothiocyanato fluorescein
PubChem CID 18730
ChEBI CHEBI:37918
IUPAC Name 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC119252500
View Documents Promotion Details
Thermo Scientific Chemicals
119252500
250 mg Glass bottle
Each for $137.85
Only null left
 
AC119250010
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
119250010
1 g Glass bottle
Each for $394.67
Only null left
 
AC119250050
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
119250050
5 g Glass bottle
Each for $1,769.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3326-32-7
389.381
MHMNJMPURVTYEJ-UHFFFAOYSA-N
18730
3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
C21H11NO5S
MFCD00005063
5-FITC, 5-Isothiocyanato fluorescein
CHEBI:37918
C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

3326-32-7
C21H11NO5S
15, 4120
MHMNJMPURVTYEJ-UHFFFAOYSA-N
Authentic
389.381
CHEBI:37918
≥88% (HPLC)
White
Fluorescein isothiocyanate isomer I
90%
MFCD00005063
5-FITC, 5-Isothiocyanato fluorescein
C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
18730
389.38
Pure
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only