missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein Isothiocyanate Isomer I, 90%, Pure

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol

Supplier:  Thermo Scientific Chemicals 119250050

Quantity 5 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC119250050

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3326-32-7
389.381
MHMNJMPURVTYEJ-UHFFFAOYSA-N
18730
3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
C21H11NO5S
MFCD00005063
5-FITC, 5-Isothiocyanato fluorescein
CHEBI:37918
C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Fluorescein isothiocyanate isomer I
90%
MFCD00005063
5-FITC, 5-Isothiocyanato fluorescein
C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
18730
389.38
Pure
White
Crystals
3326-32-7
C21H11NO5S
15, 4120
MHMNJMPURVTYEJ-UHFFFAOYSA-N
Authentic
389.381
CHEBI:37918
≥88% (HPLC)
Glass bottle
5 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions