missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein, disodium salt, pure

CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol

$70.21 - $946.69

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC173241000
View Documents Promotion Details
Thermo Scientific Chemicals
173241000
100 g Glass Bottle
Each for $70.21
Only null left
 
AC173245000
View Documents Promotion Details
Thermo Scientific Chemicals
173245000
500 g Glass Bottle
Each for $214.62
Only null left
 
AC173240025
View Documents Promotion Details
Thermo Scientific Chemicals
173240025
2.5 kg Plastic drum
Each for $946.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

518-47-8
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
C20H10Na2O5
MFCD00167039
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate

Specifications

1700 (at lambda max.) min.
518-47-8
C20H10Na2O5
19, V,6, 456
Acid Yellow 73, Fluorescein, water soluble, Uranine, C.I. 45350
NJDNXYGOVLYJHP-UHFFFAOYSA-L
Authentic
376.28
376.27
Pure
Powder
320.0°C
1.2 to 1.7
MFCD00167039
15, 4192
Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
488 to 491nm (in buffer pH 8)
10% (105°C) max.
Brown to Orange/Red or Brown/Green
Fluorescein, disodium salt
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Irritating to eyes. May cause skin and respiratory tract irritation.
NFPA
Health: 2
Flammability:

Warning

SDS
Documents

Documents

RUO – Research Use Only