missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fluorescein, disodium salt, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Absorbance | 1700 (at lambda max.) min. |
| Melting Point | 320.0°C |
| Absorption Ratio | 1.2 to 1.7 |
| Infrared Spectrum | Authentic |
| Lambda Max | 488 to 491nm (in buffer pH 8) |
| Loss on Drying | 10% (105°C) max. |
| Packaging | Glass Bottle |
| Beilstein | 19, V,6, 456 |
| Merck Index | 15, 4192 |
| Color | Brown to Orange/Red or Brown/Green |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Precautionary Statement | Emergency Overview Irritating to eyes. May cause skin and respiratory tract irritation. NFPA Health: 2 Flammability: |
RUO – Research Use Only