missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein, disodium salt, pure

CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol

Supplier:  Thermo Scientific Chemicals 173245000

 View more versions of this product

Catalog No. AC173245000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

1700 (at lambda max.) min.
320.0°C
1.2 to 1.7
MFCD00167039
15, 4192
Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Authentic
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
488 to 491nm (in buffer pH 8)
376.27
Glass Bottle
500 g
Fluorescein, disodium salt
518-47-8
C20H10Na2O5
19, V,6, 456
Acid Yellow 73, Fluorescein, water soluble, Uranine, C.I. 45350
NJDNXYGOVLYJHP-UHFFFAOYSA-L
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
376.28
10% (105°C) max.
Pure
Brown to Orange/Red or Brown/Green
Powder

Chemical Identifiers

518-47-8
376.28
NJDNXYGOVLYJHP-UHFFFAOYSA-L
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
C20H10Na2O5
MFCD00167039
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Emergency Overview
Irritating to eyes. May cause skin and respiratory tract irritation.
NFPA
Health: 2
Flammability:

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only