missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Diacetate 98.0+%, TCI America™

Catalog No. f02405g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02405G Supplier TCI America Supplier No. F02405G
Only null left

Chemical Identifiers

CAS 596-09-8
Molecular Formula C24H16O7
Molecular Weight (g/mol) 416.385
MDL Number MFCD00005062
InChI Key CHADEQDQBURGHL-UHFFFAOYSA-N
PubChem CID 65047
IUPAC Name (6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3

Specifications

Melting Point 206°C
Formula Weight 416.39
Percent Purity ≥98.0% (HPLC)
Color White/Yellow
Quantity 5 g
Physical Form Crystal/Powder
Chemical Name or Material Fluorescein Diacetate