accessibility menu, dialog, popup

UserName

CAS RN 596-09-8

IUPAC Name: 6'-(acetyloxy)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-3'-yl acetate
Synonyms: fluorescein, diacetate
Molecular Weight (g/mol): 416.39
Molecular Formula: C24H16O7
InChi Key: CHADEQDQBURGHL-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C2C(OC3=CC(OC(C)=O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)