missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein biotin, 5-((N-(5-(N-(6-(biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl)fluorescein

Catalog No. B1370
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1370 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1370 Supplier Invitrogen™ Supplier No. B1370

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-((N-(5-(N-(6-(biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl)fluorescein (fluorescein biotin)

Fluorescein biotin was developed as an alternative to radioactive biotin for detecting and quantitating biotin-binding sites by either fluorescence or absorbance. Upon binding to avidin or streptavidin, the fluorescence or absorbance of this molecule is quenched.

Specifications

Quantity 5 mg
Product Type Molecular Probes Fluorescein biotin
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Label or Dye FITC (Fluorescein)

For Research Use Only. Not for use in diagnostic procedures.