missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein 6-Isothiocyanate (isomer II) 97.0+%, TCI America™

Catalog No. F0783100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0783100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0783100MG Supplier TCI America Supplier No. F0783100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18861-78-4
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.381
MDL Number MFCD00041838
InChI Key GTQFZXYECNSNNC-UHFFFAOYSA-N
Synonym 6-Isothiocyanatofluorescein, Fluorescein Isothiocyanate, 6-FITC
PubChem CID 2762652
ChEBI CHEBI:37928
IUPAC Name 3',6'-dihydroxy-5-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N=C=S)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 389.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein 6-Isothiocyanate (isomer II)
TSCA No
Recommended Storage Refrigerator