missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, MilliporeSigma™ Supelco™

Catalog No. 111019787
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019787 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019787 Supplier Merck Emd Millipore Supplier No. 48568
Only null left
Environmental

Specifications

Packaging Ampule
CAS 86-73-7
Flash Point 151°C
Physical Form Neat
Linear Formula C13H10
MDL Number MFCD00001111
Melting Point 111°C to 114°C
Boiling Point 298°C
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 166.22
Grade Analytical Standard
Show More Show Less