Learn More
Fluorene, 98+%
CAS: 86-73-7 | C13H10 | 166.22 g/mol
Supplier: Thermo Scientific Chemicals L0212422
Description
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes
Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Fluorene | |
| 113°C to 117°C | |
| 295°C | |
| C13H10 | |
| 100 g | |
| 1363491 | |
| fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
| NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
| 9H-fluorene | |
| 6853 | |
| 166.22 |
| 86-73-7 | |
| 1.202 | |
| 151°C (303°F) | |
| MFCD00001111 | |
| UN3077 | |
| 14,4155 | |
| Soluble in hot alcohol,ether,benzene and glacial acetic acid. Insoluble in water. | |
| C1C2=CC=CC=C2C2=CC=CC=C12 | |
| 166.22 | |
| CHEBI:28266 | |
| ≥98% |
Chemical Identifiers
| 86-73-7 | |
| 166.22 | |
| NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
| 6853 | |
| 9H-fluorene |
| C13H10 | |
| MFCD00001111 | |
| fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
| CHEBI:28266 | |
| C1C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 201-695-5
RTECSNumber : LL5670000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only