missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, 98%, Spectrum™ Chemical

Catalog No. p-7165591
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
18610597 100 g Poly Bottle
1 options

Catalog No. 18610597

Supplier: Spectrum Chemical Mfg Cor FL112100GM

Only null left

Chemical Identifiers

CAS 86-73-7

Specifications

CAS Max % 1
Boiling Point 295°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.