missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, 98%, Spectrum™ Chemical

Catalog No. 18610597
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610597 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18610597 Supplier Spectrum Chemical Mfg Cor Supplier No. FL112100GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-73-7

Specifications

CAS Max % 1
Boiling Point 295°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.