missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, 98+%

Catalog No. AAL0212436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0212436 500 g
AAL0212414 25 g
AAL0212422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0212436 Supplier Thermo Scientific Chemicals Supplier No. L0212436
Only null left

CAS: 86-73-7 | C13H10 | 166.22 g/mol

Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 86-73-7
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
Synonym fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 113°C to 117°C
Density 1.202
Boiling Point 295°C
Flash Point 151°C (303°F)
Quantity 500 g
UN Number UN3077
Beilstein 1363491
Merck Index 14,4155
Solubility Information Soluble in hot alcohol,ether,benzene and glacial acetic acid. Insoluble in water.
Formula Weight 166.22
Percent Purity ≥98%
Chemical Name or Material Fluorene
Show More Show Less
Hazard Category H500
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 201-695-5
RTECSNumber LL5670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only