missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorene-2-carboxaldehyde, 99%
CAS: 30084-90-3 | C14H10O | 194.23 g/mol
Supplier: Thermo Scientific Chemicals A1491114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Fluorene-2-carboxaldehyde | |
| 82°C to 86°C | |
| MFCD00001123 | |
| 2255852 | |
| 2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, # | |
| O=CC1=CC=C2C(CC3=CC=CC=C23)=C1 | |
| 194.23 | |
| 194.23 |
| 30084-90-3 | |
| C14H10O | |
| 25 g | |
| Air Sensitive | |
| MNQGEQSXFDKAPY-UHFFFAOYSA-N | |
| 9H-fluorene-2-carbaldehyde | |
| 34804 | |
| 99% |
Chemical Identifiers
| 30084-90-3 | |
| 194.23 | |
| MNQGEQSXFDKAPY-UHFFFAOYSA-N | |
| 34804 | |
| O=CC1=CC=C2C(CC3=CC=CC=C23)=C1 |
| C14H10O | |
| MFCD00001123 | |
| 2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, # | |
| 9H-fluorene-2-carbaldehyde |
Safety and Handling
H500
EINECSNumber : 250-035-2
RTECSNumber : LL5917000
TSCA : No
RUO – Research Use Only