missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-2-carboxaldehyde, 99%

Catalog No. AAA1491122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1491122 100 g
AAA1491106 5 g
AAA1491114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1491122 Supplier Thermo Scientific Chemicals Supplier No. A1491122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30084-90-3 | C14H10O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30084-90-3
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001123
InChI Key MNQGEQSXFDKAPY-UHFFFAOYSA-N
Synonym 2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, #
PubChem CID 34804
IUPAC Name 9H-fluorene-2-carbaldehyde
SMILES O=CC1=CC=C2C(CC3=CC=CC=C23)=C1

Specifications

Melting Point 82°C to 86°C
Quantity 100 g
Beilstein 2255852
Sensitivity Air Sensitive
Formula Weight 194.23
Percent Purity 99%
Chemical Name or Material Fluorene-2-carboxaldehyde
Hazard Category H500
EINECSNumber 250-035-2
RTECSNumber LL5917000
TSCA No

RUO – Research Use Only