missing translation for 'onlineSavingsMsg'
Learn More

Fluocinolone Acetonide 98.0+%, TCI America™

$374.68 - $1247.05

Chemical Identifiers

CAS 67-73-2
Molecular Formula C24H30F2O6
Molecular Weight (g/mol) 452.50
MDL Number MFCD00010525
InChI Key FEBLZLNTKCEFIT-VSXGLTOVSA-N
Synonym fluocinolone acetonide, synalar, flucinar, synandone, fluonid, jellin, synamol, synemol, coriphate, fluovitif
PubChem CID 6215
ChEBI CHEBI:31623
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F06571G
View Documents
TCI America
F06571G
1 g
Each for $374.68
Only null left
 
F06575G
View Documents
TCI America
F06575G
5 g
Each for $1,247.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-73-2
452.50
FEBLZLNTKCEFIT-VSXGLTOVSA-N
6215
(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
C24H30F2O6
MFCD00010525
fluocinolone acetonide, synalar, flucinar, synandone, fluonid, jellin, synamol, synemol, coriphate, fluovitif
CHEBI:31623
CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

67-73-2
White-Yellow
MFCD00010525
fluocinolone acetonide, synalar, flucinar, synandone, fluonid, jellin, synamol, synemol, coriphate, fluovitif
CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
452.50
CHEBI:31623
≥98.0% (HPLC)
Fluocinolone Acetonide
266°C
C24H30F2O6
1 g
FEBLZLNTKCEFIT-VSXGLTOVSA-N
(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
6215
452.49
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TU3830000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'