Learn More
Molecular Probes™ Fluo-4, Pentapotassium Salt, cell impermeant
Description
Fluo-4, Pentapotassium Salt, cell impermeant
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be physically loaded with the cell-impermeant salt forms of these indicators using patch pipette, microinjection, or our Influx™ pinocytotic cell-loading reagent (Cat. No. I14402). These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.
Calcium Indicator (Cell-Impermeant Salts) Specifications:
- Label (Ex/Em of Ca2+–bound form): Fluo-4 (494/516 nm)
- Fluorescence intensity increase upon binding Ca2+: >100 fold
- Kd for Ca2+ in buffer: ∼335 nM
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
Using TPEN to Control Heavy Metal Cations
In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator (Cat. No. TPEN).
More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.
For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Indicator |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager |
| Product Type | Calcium Indicator |
| Dye Type | Fluo-4 |
| Excitation Wavelength Range | 480 to 495 nm |
| Quantity | 500 μg |
| Shipping Condition | Room Temperature |
| Emission Wavelength Range | 510 to 525 nm |
| Show More |
Frequently Asked Questions (FAQs)
No. pH affects the ability of Fluo-4, Pentapotassium Salt, cell impermeant (Cat. No. F14200) to bind calcium and the fluorescence intensity. The Fluo-4 dye drops in fluorescence below pH 5.
For Research Use Only. Not for use in diagnostic procedures.