missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fluo-4 NW Calcium Assay Kit
Description
Fluo-4 NW Calcium Assay Kit
*The Fluo-4 NW (no-wash) Calcium Assay Kits provide a simplified method for detecting intracellular calcium, without a quencher dye or a wash step.
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Assay |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Stain |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 10 Microplates |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.