Learn More
Molecular Probes™ Fluo-4 Direct™ Calcium Assay Kit
Description
Fluo-4 Direct™ Calcium Assay Kit
The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:
- Easily loads into cells
- Achieves a large assay window and
- Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence
This advanced formulation allows the assay to be run in a simple 'addition only' format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.
The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:
- Easily loads into cells
- Achieves a large assay window and
- Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence
This advanced formulation allows the assay to be run in a simple 'addition only' format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.
Specifications
Specifications
| Color | Green |
| Content And Storage |
|
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Indicator |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager |
| Product Type | Calcium Assay Kit |
| Dye Type | Fluo-4 Direct |
| Excitation Wavelength Range | 480 to 495 nm, 495 to 510 nm |
| Form | Powder |
| Modification | Chemical |
| Show More |
Frequently Asked Questions (FAQs)
The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).
After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.
For Research Use Only. Not for use in diagnostic procedures.