missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flumequine, 98%
Inhibits topoisomerases in gram negative bacteria | CAS: 42835-25-6 | C14H12FNO3 | 261.252 g/mol
$93.56 - $278.92
Chemical Identifiers
| CAS | 42835-25-6 |
|---|---|
| Molecular Formula | C14H12FNO3 |
| Molecular Weight (g/mol) | 261.252 |
| MDL Number | MFCD00079298 |
| InChI Key | DPSPPJIUMHPXMA-UHFFFAOYSA-N |
| Synonym | 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid |
| PubChem CID | 3374 |
| ChEBI | CHEBI:85269 |
| SMILES | CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6633203
|
Thermo Scientific Chemicals
J6633203 |
1 g |
Each for $93.56
|
|
|||||
|
AAJ6633206
|
Thermo Scientific Chemicals
J6633206 |
5 g |
Each for $278.92
|
|
|||||
Description
Inhibits topoisomerases in Gram-negative bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 42835-25-6 | |
| 261.252 | |
| DPSPPJIUMHPXMA-UHFFFAOYSA-N | |
| 3374 | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O |
| C14H12FNO3 | |
| MFCD00079298 | |
| 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| CHEBI:85269 |
Specifications
| Flumequine | |
| 42835-25-6 | |
| White | |
| >110°C (230°F) | |
| MFCD00079298 | |
| 14,4137 | |
| Soluble in DMSO and dilute alkali hydroxides; Insoluble in water | |
| CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O | |
| 3374 | |
| 261.25 | |
| Flumequine |
| Crystalline Powder | |
| 253°C to 255°C | |
| 1 g | |
| C14H12FNO3 | |
| 490724 | |
| 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid | |
| DPSPPJIUMHPXMA-UHFFFAOYSA-N | |
| 261.252 | |
| CHEBI:85269 | |
| 98% |
Safety and Handling
EINECSNumber : 255-962-6
RTECSNumber : DK1672000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only