missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FluidEase™ Pro ClipTip™ Electronic Single-Channel Pipettes

Catalog No. 01671143
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 300 μL
2.5 to 100 μL
20 to 1,000 μL
5 to 200 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01671143 0.5 to 10 μL
01671142 2.5 to 100 μL
01671141 5 to 200 μL
01671140 10 to 300 μL
01671139 20 to 1,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01671143 Supplier Thermo Scientific™ Supplier No. 4674010
Only null left

Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic single-channel pipettes combine a large touchscreen, ultra-wide speed selection, and secure ClipTip™ sealing to deliver precise, reproducible pipetting with faster training and confident use in shared laboratories.

Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic single-channel pipettes simplify electronic pipetting while expanding workflow control and standardization. A large, high-resolution touchscreen and ultra-wide speed selection enable intuitive setup and precise handling across liquid types. Secure ClipTip™ sealing, preset and advanced programming modes, password protection, and service and calibration logs support reproducible results and confident operation in shared and regulated laboratories.

Features:

  • Large Touchscreen User Interface: A high-resolution touchscreen provides direct, visual interaction for setup and execution, reducing training time and minimizing setup errors compared to button- or joystick-based navigation.
  • Ultra-Wide Speed Selection: One of the broadest aspiration and dispense speed ranges available supports gentle handling of sensitive samples, precise control of viscous liquids, and faster execution for time-critical workflows.
  • Simple Presets and Advanced Programming: Easy-to-use preset modes support fast onboarding for new or occasional users, while advanced step programming enables complex, multi-step protocols for demanding applications.
  • Index-Finger Operation with Increased Rotational Flexibility: Retains Thermo Fisher–recognized index-finger operation with a wider angle of rotation at the finger rest to accommodate different hand sizes, grip angles, and working positions.
  • Secure ClipTip™ Technology: ClipTip™ tips lock and seal in place until intentional ejection, helping ensure consistent volume delivery and reduced variability from dispense to dispense.
  • Electronic Tip Ejection with Safe-to-Eject Illumination: Electronic tip ejection reduces physical effort and includes LED illumination to indicate when ejection is possible, providing ergonomic benefit and added user security.
  • Password Protection and User Profiles: Password-protect critical settings and protocols and support up to five user profiles to enable standardized operation in shared and regulated laboratory environments.
  • Custom Calibration Options: Enable calibration to customer-specific liquids or environmental conditions to align performance with real laboratory workflows.
  • Service and Maintenance Tracking: Built-in service reminders, calibration logs, and usage tracking support instrument management and compliance documentation over time.

Specifications

Compatible Tips ClipTip 12.5
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Pink
Disposable Reusable
For Use With ClipTip 12.5
Model Bluetooth
Number of Channels 1
Programmability Yes
Sterility Non-sterile
Type Electronic Single Channel Pipette
Volume (Metric) 0.5 to 10 μL
Display Touchscreen
Finger Style Ergonomic Index Finger
Multilingual User Interface Yes
Warranty 1 Year, 2 Years with Registration
Show More Show Less