missing translation for 'onlineSavingsMsg'
Learn More

Flufenamic acid, MilliporeSigma™ Supelco™

Catalog No. 111020607
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020607 10 g
111019379 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020607 Supplier Merck Emd Millipore Supplier No. F900510G

Flufenamic acid, a non-steroidal anti-inflammatory drug, is an effective analgesic agent.

Pharmaceutical (small molecule)

Specifications

CAS 530-78-9
Physical Form Neat
Linear Formula 2-(CF3C6H4 NH)C6H4 CO2 H
MDL Number MFCD00002422
Melting Point 132°C to 135°C
Synonym N-(3-[Trifluoromethyl]phenyl)anthranilic acid; N-(alpha,alpha,alpha-Trifluoro-m-tolyl)anthranilic acid
Type Analytical Standard
Industry Type Drug Analysis
Quantity 10 g
Formula Weight 281.23
Grade Analytical Standard
Show More Show Less