missing translation for 'onlineSavingsMsg'
Learn More

FLT3LG Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

$337.00 - $432.00

Specifications

Antigen FLT3LG
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
500P42BT25 DFS Item
View Documents
PeproTech
500P42BT25UG
25 μg
Each for $337.00
Only null left
 
500P42BT50 DFS Item
View Documents
PeproTech
500P42BT50UG
50 μg
Each for $432.00
Only null left
 
50-271-8487
View Documents
PeproTech
500P42BT500UG
500 μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

AA Sequence of recombinant protein: TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVASNLQD EELCGGLWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQPP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPWSPRPLEA TAPTA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Flt3-ligand. Anti-Human Flt3-ligand-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Flt3-ligand by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Flt3-ligand (500-P42) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Flt3-ligand. Western Blot: To detect Human Flt3-ligand by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Flt3-ligand is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P42BT-1MG will be provided as 2 x 500 μg

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
Specifications

Specifications

FLT3LG
ELISA, Western Blot
Biotin
Rabbit
Human
P49771
2323
E.coli-derived Recombinant Human Flt3-Ligand.
Antigen affinity chromatography
FLT3LG
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
FLT3LG
Primary
-20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'