missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLT3LG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149799
Description
FLT3LG Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.Specifications
| FLT3LG | |
| Polyclonal | |
| Unconjugated | |
| Flt3l | |
| FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine | |
| Rabbit | |
| Protein A | |
| RUO | |
| 103691134, 14256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P49772 | |
| Flt3l, FLT3LG | |
| Recombinant FMS Like Tyrosine Kinase 3 Ligand (Flt3L) (Gly27-Gln189). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |