missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FLRT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5150063
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5150063 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5150063 Supplier Invitrogen™ Supplier No. PA5150063
Only null left

Rabbit Polyclonal Antibody

FLRT2 Polyclonal Antibody for IHC (P)

This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRT family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix.

Specifications

Antigen FLRT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FLRT2
Gene Accession No. O43155
Gene Alias Fibronectin leucine rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2; FLRT2; Kiaa0405; Leucine-rich repeat transmembrane protein FLRT2; UNQ232/PRO265
Gene Symbols FLRT2
Host Species Rabbit
Immunogen Recombinant Fibronectin Leucine Rich Transmembrane Protein 2 (FLRT2) (Ser300-Ser517).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23768
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less