Learn More
Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative. Designed for use in immunofluorescent staining protocols of cells in suspension.
Supplier:  Invitrogen™ 00422257
 View more versions of this product
Catalog No. 501128916
Only null left
Alert:
Contains Sodium azide
Description
Description
  • Designed for use in immunofluorescent staining protocols of cells in suspension
  • A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative
  • This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
  • Contains animal serum proteins to help minimizing non-specific binding of antibodies
Specifications
Chemical Identifiers
eBioscience
Staining Buffer
Specifications
200 mL
eBioscience
Staining Buffer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.