Learn More
Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
Description
- Designed for use in immunofluorescent staining protocols of cells in suspension
- A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative
- This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
- Contains animal serum proteins to help minimizing non-specific binding of antibodies
Specifications
Specifications
| Quantity | 600 mL |
| Product Line | eBioscience |
| Product Type | Staining Buffer |
For Research Use Only. Not for use in diagnostic procedures.